Assay Details
Gene Symbol: Tmem98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007672.1 | NP_001007673.1 | 1 - 2 | 70 | 108 | |
GenBank mRNA | BC078932.1 | - | 1 - 2 | 70 | 108 | |
FQ214419.1 | - | 1 - 2 | 89 | 108 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC078932.1, NM_001007672.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
