Assay Details
Gene Symbol: Tmem98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007672.1 | NP_001007673.1 | 3 - 4 | 392 | 132 | |
GenBank mRNA | BC078932.1 | - | 3 - 4 | 392 | 132 | |
FQ214419.1 | - | 3 - 4 | 411 | 267 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
