Assay Details
Gene Symbol: Mlx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034112.1 | NP_001029284.1 | 1 - 2 | 214 | 114 | |
GenBank mRNA | BC128702.1 | - | 1 - 2 | 248 | 114 | |
CO560831.1 | - | 1 - 2 | 214 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
