Assay Details
Gene Symbol: Sphk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270807.1 | NP_001257736.1 | 2 - 3 | 501 | 80 | |
NM_001270808.1 | NP_001257737.1 | 2 - 3 | 550 | 80 | ||
NM_001270809.1 | NP_001257738.1 | 2 - 3 | 451 | 80 | ||
NM_001270810.1 | NP_001257739.1 | 2 - 3 | 1137 | 80 | ||
NM_001270811.1 | NP_001257740.1 | 2 - 3 | 1487 | 80 | ||
NM_133386.3 | NP_596877.2 | 1 - 2 | 1741 | 80 | ||
XM_006247788.2 | XP_006247850.1 | 2 - 3 | 332 | 80 | ||
GenBank mRNA | AB049571.1 | - | 2 - 3 | 549 | 80 | |
AB049572.1 | - | 2 - 3 | 501 | 80 | ||
AB049573.1 | - | 2 - 3 | 448 | 80 | ||
AB049574.1 | - | 2 - 3 | 1107 | 80 | ||
AB049575.1 | - | 1 - 2 | 1254 | 80 | ||
BC081738.1 | - | 1 - 2 | 1706 | 7132896 | 80 | |
DQ486894.1 | - | 2 - 3 | 289 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB049571.1, AB049572.1, AB049573.1, AB049574.1, AB049575.1, BC081738.1, NM_001270807.1, NM_001270808.1, NM_001270809.1, NM_001270810.1, NM_133386.3, XM_006247788.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
