Assay Details
Gene Symbol: Mlst8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022404.2 | NP_071799.2 | 4 - 5 | 703 | 83 | |
GenBank mRNA | AF051155.2 | - | 4 - 5 | 702 | 83 | |
BC097319.1 | - | 4 - 5 | 381 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
