Assay Details
Gene Symbol: Ppp1r1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138521.1 | NP_612530.1 | 1 - 2 | 503 | 116 | |
GenBank mRNA | AY601872.1 | - | 1 - 2 | 503 | 116 | |
BC078954.1 | - | 1 - 2 | 433 | 7110316 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
