Assay Details
Gene Symbol: Skap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173311.2 | NP_775433.2 | 5 - 6 | 455 | 96 | |
GenBank mRNA | AB092812.1 | - | 5 - 6 | 357 | 96 | |
BC097931.1 | - | 5 - 6 | 455 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
