Assay Details
Gene Symbol: Pcdh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251023.2 | XP_006251085.1 | 1 - 2 | 3258 | 75 | |
GenBank mRNA | AY690613.1 | - | 1 - 2 | 3180 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
