Assay Details
Gene Symbol: Tbrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012154.1 | NP_001012154.1 | 8 - 9 | 1721 | 89 | |
XM_006251342.2 | XP_006251404.1 | 10 - 11 | 1903 | 89 | ||
XM_006251343.2 | XP_006251405.1 | 10 - 11 | 1833 | 89 | ||
GenBank mRNA | BC087073.1 | - | 8 - 9 | 1721 | 89 | |
FQ217643.1 | - | 10 - 11 | 1866 | 89 | ||
FQ217828.1 | - | 10 - 11 | 1866 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
