Assay Details
Gene Symbol: Tbrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012154.1 | NP_001012154.1 | 9 - 10 | 1833 | 78 | |
XM_006251342.2 | XP_006251404.1 | 11 - 12 | 2015 | 78 | ||
XM_006251343.2 | XP_006251405.1 | 11 - 12 | 1945 | 78 | ||
GenBank mRNA | BC087073.1 | - | 9 - 10 | 1833 | 7134456 | 78 |
FQ217643.1 | - | 11 - 12 | 1978 | 78 | ||
FQ217828.1 | - | 11 - 12 | 1978 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
