Assay Details
Gene Symbol: Tbrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012154.1 | NP_001012154.1 | 2 - 3 | 777 | 80 | |
XM_006251342.2 | XP_006251404.1 | 4 - 5 | 959 | 80 | ||
XM_006251343.2 | XP_006251405.1 | 4 - 5 | 889 | 80 | ||
GenBank mRNA | BC087073.1 | - | 2 - 3 | 777 | 7134456 | 80 |
FQ217643.1 | - | 4 - 5 | 922 | 80 | ||
FQ217828.1 | - | 4 - 5 | 922 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
