Assay Details
Gene Symbol: Oxnad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107295.1 | NP_001100765.1 | 4 - 5 | 491 | 154 | |
XM_006252623.2 | XP_006252685.1 | 4 - 5 | 592 | 154 | ||
XM_006252624.2 | XP_006252686.1 | 5 - 6 | 699 | 154 | ||
XM_006252625.2 | XP_006252687.1 | 5 - 6 | 685 | 154 | ||
XM_006252626.2 | XP_006252688.1 | 4 - 5 | 578 | 154 | ||
XM_008771000.1 | XP_008769222.1 | 5 - 6 | 780 | 154 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
