Assay Details
Gene Symbol: Glud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012570.2 | NP_036702.1 | 9 - 10 | 1442 | 83 | |
GenBank mRNA | BC081841.1 | - | 9 - 10 | 1326 | 83 | |
X14044.1 | - | 9 - 10 | 1332 | 83 | ||
X14223.1 | - | 9 - 10 | 1305 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
