Assay Details
Gene Symbol: Mtus2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100989.1 | NP_001094459.1 | 1 - 2 | 124 | 87 | |
GenBank mRNA | BC127538.1 | - | 1 - 2 | 124 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
