Assay Details
Gene Symbol: Mtus2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100989.1 | NP_001094459.1 | 7 - 8 | 803 | 99 | |
XM_008768993.1 | XP_008767215.1 | 11 - 12 | 1708 | 99 | ||
XM_008768994.1 | XP_008767216.1 | 11 - 12 | 1562 | 99 | ||
GenBank mRNA | BC127538.1 | - | 7 - 8 | 803 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P18464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9284 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P18464 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P18464 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P18464 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P18464 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P395918 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 8839 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10760028 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10760032 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10760027 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10760030 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10760033 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10760034 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10760031 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10760026 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10760025 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10760024 | - | - |
Rat Genome 230 2.0 Array | 1390051_at | - | - |
Rat Genome 230 2.0 Array | 1390051_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390051_at | - | - |
Affymetrix HT Rat Focus Array | 1390051_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1200008737.rn.1 | TC1200001027.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200008736.rn.1 | TC1200001027.rn.1 | RTA TC |
Rat Expression Set 230 | 1390051_PM_at | - | - |
Rat Expression Set 230 | 1390051_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10760024 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21483361 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367982 | - | - |
