Assay Details
Gene Symbol: Get4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163320.1 | NP_001156792.1 | 3 - 4 | 429 | 101 | |
XM_006248927.2 | XP_006248989.1 | 3 - 4 | 2665 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
