Assay Details
Gene Symbol: Lhpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009706.1 | NP_001009706.1 | 6 - 7 | 726 | 78 | |
XM_008759912.1 | XP_008758134.1 | 6 - 7 | 2111 | 78 | ||
GenBank mRNA | BC088448.1 | - | 6 - 7 | 726 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P445444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826473 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671605 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952620 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80713 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P445444 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P445444 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P445444 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P445444 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 63322 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49873 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 80630 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 51597 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10711722 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10711717 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10711719 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10711720 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10711721 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10711718 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10711716 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10711715 | - | - |
Rat Genome 230 2.0 Array | 1372860_PM_at | - | - |
Rat Genome 230 2.0 Array | 1372860_at | - | - |
Affymetrix HT Rat Focus Array | 1372860_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372860_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100021672.rn.1 | TC0100002750.rn.1 | RTA TC |
Rat Expression Set 230 | 1372860_at | - | - |
Rat Expression Set 230 | 1372860_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10711715 | - | - |
Rat Genome U34 Set | rc_AI233941_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22320018 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1530391 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357051 | - | - |
RatRef-12 Expression BeadChip | ILMN_66014 | - | - |
RatRef-12 Expression BeadChip | ILMN_160901 | - | - |
