Assay Details
Gene Symbol: Uros | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012068.1 | NP_001012068.1 | 6 - 7 | 550 | 116 | |
XM_008759890.1 | XP_008758112.1 | 5 - 6 | 751 | 116 | ||
XM_008759891.1 | XP_008758113.1 | 5 - 6 | 446 | 116 | ||
XM_008759892.1 | XP_008758114.1 | 5 - 6 | 719 | 116 | ||
GenBank mRNA | BC083730.1 | - | 6 - 7 | 550 | 7191339 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
