Assay Details
Gene Symbol: Lgi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145769.3 | NP_665712.1 | 2 - 3 | 283 | 87 | |
GenBank mRNA | AJ487517.1 | - | 2 - 3 | 283 | 87 | |
BC089222.1 | - | 2 - 3 | 390 | 7320899 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
