Assay Details
Gene Symbol: Tmem132a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178021.1 | NP_821140.1 | 9 - 10 | 1824 | 62 | |
XM_006231041.2 | XP_006231103.1 | 9 - 10 | 2762 | 62 | ||
GenBank mRNA | AY216677.1 | - | 9 - 10 | 1824 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
