Assay Details
Gene Symbol: Sipa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004089.1 | NP_001004089.1 | 7 - 8 | 2021 | 68 | |
XM_006230849.2 | XP_006230911.1 | 8 - 9 | 2638 | 68 | ||
GenBank mRNA | AB091474.1 | - | 7 - 8 | 2021 | 68 | |
FQ223350.1 | - | 8 - 9 | 2228 | 68 | ||
FQ228273.1 | - | 8 - 9 | 2207 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
