Assay Details
Gene Symbol: Dagla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005886.1 | NP_001005886.1 | 13 - 14 | 1456 | 79 | |
XM_008760229.1 | XP_008758451.1 | 12 - 13 | 4826 | 79 | ||
XM_008760230.1 | XP_008758452.1 | 13 - 14 | 4741 | 79 | ||
GenBank mRNA | AY275375.1 | - | 13 - 14 | 1456 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
