Assay Details
Gene Symbol: Stip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138911.3 | NP_620266.1 | 11 - 12 | 1347 | 67 | |
GenBank mRNA | BC061529.1 | - | 11 - 12 | 1348 | 67 | |
FQ220758.1 | - | 8 - 9 | 869 | 67 | ||
FQ227512.1 | - | 2 - 3 | 98 | 67 | ||
Y15068.1 | - | 10 - 11 | 1282 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
