Assay Details
Gene Symbol: Stip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138911.3 | NP_620266.1 | 4 - 5 | 566 | 77 | |
GenBank mRNA | BC061529.1 | - | 4 - 5 | 567 | 6887497 | 77 |
FQ220758.1 | - | 1 - 2 | 88 | 77 | ||
Y15068.1 | - | 3 - 4 | 501 | 77 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
