Assay Details
Gene Symbol: Stip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138911.3 | NP_620266.1 | 7 - 8 | 963 | 158 | |
GenBank mRNA | BC061529.1 | - | 7 - 8 | 964 | 6887497 | 158 |
FQ220758.1 | - | 4 - 5 | 485 | 158 | ||
Y15068.1 | - | 6 - 7 | 898 | 158 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
