Assay Details
Gene Symbol: Lsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025420.1 | NP_001020591.1 | 5 - 6 | 611 | 92 | |
XM_006230815.2 | XP_006230877.1 | 6 - 7 | 1273 | 92 | ||
XM_006230817.2 | XP_006230879.1 | 5 - 6 | 648 | 92 | ||
XM_006230818.1 | XP_006230880.1 | 5 - 6 | 706 | 92 | ||
XM_008760146.1 | XP_008758368.1 | 5 - 6 | 686 | 92 | ||
XM_008760147.1 | XP_008758369.1 | 5 - 6 | 935 | 92 | ||
GenBank mRNA | BC097966.1 | - | 5 - 6 | 585 | 92 | |
FQ230230.1 | - | 5 - 6 | 592 | 92 | ||
FQ232610.1 | - | 5 - 6 | 601 | 92 | ||
FQ233963.1 | - | 5 - 6 | 601 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
