Assay Details
Gene Symbol: Lsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025420.1 | NP_001020591.1 | 6 - 7 | 650 | 73 | |
XM_006230815.2 | XP_006230877.1 | 7 - 8 | 1312 | 73 | ||
XM_006230817.2 | XP_006230879.1 | 6 - 7 | 687 | 73 | ||
XM_006230818.1 | XP_006230880.1 | 6 - 7 | 745 | 73 | ||
XM_008760146.1 | XP_008758368.1 | 6 - 7 | 725 | 73 | ||
XM_008760147.1 | XP_008758369.1 | 6 - 7 | 974 | 73 | ||
GenBank mRNA | BC097966.1 | - | 6 - 7 | 624 | 73 | |
FQ232610.1 | - | 6 - 7 | 640 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
