Assay Details
Gene Symbol: Lsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025420.1 | NP_001020591.1 | 7 - 8 | 730 | 84 | |
XM_006230815.2 | XP_006230877.1 | 8 - 9 | 1392 | 84 | ||
XM_006230817.2 | XP_006230879.1 | 7 - 8 | 767 | 84 | ||
XM_006230818.1 | XP_006230880.1 | 7 - 8 | 825 | 84 | ||
XM_008760146.1 | XP_008758368.1 | 7 - 8 | 805 | 84 | ||
XM_008760147.1 | XP_008758369.1 | 7 - 8 | 1054 | 84 | ||
GenBank mRNA | BC097966.1 | - | 7 - 8 | 704 | 84 | |
FQ230230.1 | - | 7 - 8 | 711 | 84 | ||
FQ232610.1 | - | 7 - 8 | 720 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
