Assay Details
Gene Symbol: Pank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106373.3 | NP_001099843.1 | 1 - 2 | 200 | 79 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001106373.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
