Assay Details
Gene Symbol: Pank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106373.3 | NP_001099843.1 | 2 - 3 | 557 | 122 | |
XM_006231276.2 | XP_006231338.1 | 2 - 3 | 3860 | 122 | ||
XM_006231277.2 | XP_006231339.1 | 2 - 3 | 3865 | 122 | ||
XM_006231278.2 | XP_006231340.1 | 3 - 4 | 1208 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
