Assay Details
Gene Symbol: Snx15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024752.1 | NP_001019923.1 | 1 - 2 | 190 | 96 | |
GenBank mRNA | BC097481.1 | - | 1 - 2 | 190 | 7459091 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
