Assay Details
Gene Symbol: Cd151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230576.2 | XP_006230638.1 | 191 | 82 |
Gene Symbol: LOC100911730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230598.2 | XP_006230660.1 | 1 - 2 | 191 | 82 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the XM_006230576.2,XM_006230598.2 transcripts.
This assay is in the 5'UTR region of the XM_006230576.2,XM_006230598.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
