Assay Details
Gene Symbol: Eef1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004223.2 | NP_001004223.1 | 4 - 5 | 514 | 77 | |
GenBank mRNA | BC079398.1 | - | 5 - 6 | 523 | 7112627 | 77 |
BC098895.1 | - | 4 - 5 | 514 | 7367246 | 77 | |
FQ212596.1 | - | 5 - 6 | 532 | 77 | ||
FQ213179.1 | - | 5 - 6 | 530 | 77 | ||
FQ214210.1 | - | 5 - 6 | 529 | 77 | ||
FQ214443.1 | - | 5 - 6 | 531 | 77 | ||
FQ215853.1 | - | 5 - 6 | 560 | 77 | ||
FQ216520.1 | - | 3 - 4 | 299 | 77 | ||
FQ218274.1 | - | 5 - 6 | 530 | 77 | ||
FQ219298.1 | - | 5 - 6 | 532 | 77 | ||
FQ222930.1 | - | 5 - 6 | 530 | 77 | ||
FQ223614.1 | - | 5 - 6 | 530 | 77 | ||
FQ224956.1 | - | 4 - 5 | 489 | 77 | ||
FQ228703.1 | - | 5 - 6 | 530 | 77 | ||
FQ228918.1 | - | 5 - 6 | 531 | 77 | ||
FQ230995.1 | - | 5 - 6 | 530 | 77 | ||
FQ231923.1 | - | 5 - 6 | 530 | 77 | ||
FQ233868.1 | - | 5 - 6 | 530 | 77 | ||
FQ235034.1 | - | 5 - 6 | 680 | 77 |
Gene Symbol: Slamf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250250.2 | XP_006250312.1 | 7 | 2806 | 77 | |
GenBank mRNA | FQ230857.1 | - | 3 | 1740 | 77 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
