Assay Details
Gene Symbol: Ap2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031008.2 | NP_112270.2 | 11 - 12 | 1534 | 92 | |
XM_008760010.1 | XP_008758232.1 | 11 - 12 | 1589 | 92 | ||
GenBank mRNA | BC081786.1 | - | 11 - 12 | 1534 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
