Assay Details
Gene Symbol: Ppp1ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031527.1 | NP_113715.1 | 1 - 2 | 123 | 87 | |
GenBank mRNA | BC070517.1 | - | 1 - 2 | 103 | 87 | |
D00859.1 | - | 1 - 2 | 123 | 87 | ||
D90163.1 | - | 1 - 2 | 107 | 87 | ||
FQ224860.1 | - | 1 - 2 | 113 | 87 | ||
FQ231613.1 | - | 1 - 2 | 132 | 87 | ||
FQ232496.1 | - | 1 - 2 | 131 | 87 | ||
FQ233040.1 | - | 1 - 2 | 123 | 87 | ||
FQ235125.1 | - | 1 - 2 | 123 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
