Assay Details
Gene Symbol: Lipk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106374.1 | NP_001099844.1 | 2 - 3 | 204 | 77 | |
XM_008760325.1 | XP_008758547.1 | 3 - 4 | 212 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
