Assay Details
Gene Symbol: Hnrnpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134760.1 | NP_001128232.1 | 8 - 9 | 1460 | 90 | |
NM_032619.2 | NP_116008.2 | 9 - 10 | 1349 | 90 | ||
XM_006228691.2 | XP_006228753.1 | 8 - 9 | 1604 | 90 | ||
XM_006228692.2 | XP_006228754.1 | 9 - 10 | 1493 | 90 | ||
GenBank mRNA | AB260892.1 | - | 2 - 3 | 326 | 90 | |
BC086392.1 | - | 5 - 6 | 574 | 90 | ||
BC169048.1 | - | 9 - 10 | 1304 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
