Assay Details
Gene Symbol: Akt1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106259.3 | NP_001099729.2 | 3 - 4 | 577 | 128 | |
XM_006229003.2 | XP_006229065.1 | 3 - 4 | 1053 | 128 | ||
XM_006229004.2 | XP_006229066.1 | 3 - 4 | 753 | 128 | ||
XM_006229005.2 | XP_006229067.1 | 3 - 4 | 729 | 128 | ||
GenBank mRNA | EV773331.1 | - | 2 - 3 | 493 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
