Assay Details
Gene Symbol: Slc1a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175758.3 | NP_786934.1 | 5 - 6 | 1447 | 82 | |
GenBank mRNA | AJ132846.1 | - | 5 - 6 | 1178 | 82 | |
BC080242.1 | - | 5 - 6 | 1447 | 7100054 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
