Assay Details
Gene Symbol: Dmwd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759037.1 | XP_008757259.1 | 4 - 5 | 2025 | 99 | |
GenBank mRNA | BC160851.1 | - | 4 - 5 | 1922 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
