Assay Details
Gene Symbol: Ppp1r13l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228495.2 | XP_006228557.1 | 4 - 5 | 3599 | 136 | |
XM_008759052.1 | XP_008757274.1 | 5 - 6 | 3134 | 136 | ||
XM_008759053.1 | XP_008757275.1 | 4 - 5 | 2303 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
