Assay Details
Gene Symbol: Slc31a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133600.1 | NP_598284.1 | 2 - 3 | 291 | 72 | |
GenBank mRNA | AF268030.1 | - | 2 - 3 | 291 | 72 | |
AY539951.1 | - | 1 - 2 | 123 | 72 | ||
BC078745.1 | - | 2 - 3 | 253 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
