Assay Details
Gene Symbol: Tyrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106664.1 | NP_001100134.1 | 6 - 7 | 1507 | 85 | |
XM_006238332.2 | XP_006238394.1 | 6 - 7 | 1538 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
