Assay Details
Gene Symbol: Hcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053685.1 | NP_446137.1 | 5 - 6 | 1237 | 87 | |
GenBank mRNA | AF247452.1 | - | 5 - 6 | 1237 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
