Assay Details
Gene Symbol: Tmem144 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108551.1 | NP_001102021.1 | 4 - 5 | 676 | 96 | |
XM_006232517.2 | XP_006232579.1 | 4 - 5 | 677 | 96 | ||
GenBank mRNA | FQ211169.1 | - | 2 - 3 | 230 | 96 | |
FQ221228.1 | - | 2 - 3 | 230 | 96 | ||
FQ223804.1 | - | 2 - 3 | 230 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
