Assay Details
Gene Symbol: Tmem144 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108551.1 | NP_001102021.1 | 7 - 8 | 920 | 101 | |
XM_006232517.2 | XP_006232579.1 | 7 - 8 | 921 | 101 | ||
GenBank mRNA | FQ211169.1 | - | 5 - 6 | 474 | 101 | |
FQ223804.1 | - | 5 - 6 | 474 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
