Assay Details
Gene Symbol: Pklr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232593.2 | XP_006232655.1 | 1 - 2 | 152 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
