Assay Details
Gene Symbol: Pklr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012624.3 | NP_036756.3 | 6 - 7 | 882 | 92 | |
XM_006232593.2 | XP_006232655.1 | 6 - 7 | 1013 | 92 | ||
GenBank mRNA | BC161827.1 | - | 5 - 6 | 872 | 92 | |
M11709.1 | - | 6 - 7 | 883 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
