Assay Details
Gene Symbol: Kcnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019315.2 | NP_062188.2 | 5 - 6 | 1999 | 114 | |
GenBank mRNA | AF284345.1 | - | 5 - 6 | 1696 | 114 | |
AF292389.1 | - | 5 - 6 | 1999 | 114 | ||
U69884.1 | - | 5 - 6 | 1703 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
