Assay Details
Gene Symbol: Asic5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022227.1 | NP_071563.1 | 3 - 4 | 676 | 184 | |
GenBank mRNA | Y19034.1 | - | 3 - 4 | 676 | 184 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
